
4-(4-Chlorophenoxy)-2-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1518092-44-8
:
Formula:C₁₄H₈ClF₃O₂
MDL Number:MFCD31812101
:300.66 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together