Order Entry
Italy
ContactUsLinkComponent
1-(4-Fluorophenyl)-4-(morpholin-4-yl)butan-1-one, tech
1-(4-Fluorophenyl)-4-(morpholin-4-yl)butan-1-one, tech
Catalog # APOSPC28198-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluorophenyl)-4-(morpholin-4-yl)butan-1-one, tech
Catalog # APOSPC28198-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.297 g/mol
Storage Temperature:Ambient
CAS Number:3800-15-5
Formula:C₁₄H₁₈FNO₂
MDL Number:MFCD01737578