Order Entry
Italy
ContactUsLinkComponent
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech
Catalog # APOSPC28154-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-(4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech
Catalog # APOSPC28154-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.258 g/mol
Storage Temperature:Ambient
CAS Number:866133-01-9
Formula:C₁₅H₁₁FN₂O₂
MDL Number:MFCD04126009