Order Entry
Italy
ContactUsLinkComponent
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid, tech
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid, tech
Catalog # APOSPC28133-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluorophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid, tech
Catalog # APOSPC28133-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.232 g/mol
Storage Temperature:Ambient
CAS Number:133427-95-9
Formula:C₁₄H₉FN₂O₂
MDL Number:MFCD05256204