Order Entry
Italy
ContactUsLinkComponent
2-Hydroxy-3-phenyl-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one, tech
2-Hydroxy-3-phenyl-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one, tech
Catalog # APOSPC28127-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Hydroxy-3-phenyl-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one, tech
Catalog # APOSPC28127-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.181 g/mol
Storage Temperature:Ambient
CAS Number:26676-02-8
Formula:C₁₁H₇F₃N₂O₂
MDL Number:MFCD03102547