1,1,1-Trifluoro-3-(4-phenylpiperazin-1-yl)propan-2-ol, tech
Catalog # APOSPC28122-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:274.282 g/mol
Storage Temperature:Ambient
CAS Number:672952-04-4
Formula:C₁₃H₁₇F₃N₂O
MDL Number:MFCD02083036