Order Entry
Italy
ContactUsLinkComponent
Ethyl 8-chloro-6-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Ethyl 8-chloro-6-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Catalog # APOSPC28116-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 8-chloro-6-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Catalog # APOSPC28116-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.656 g/mol
Storage Temperature:Ambient
CAS Number:338795-20-3
Formula:C₁₂H₉ClFNO₃
MDL Number:MFCD00173402