Order Entry
Italy
ContactUsLinkComponent
Methyl 2,2-difluoro-2-(4-fluorophenoxy)acetate, tech
Methyl 2,2-difluoro-2-(4-fluorophenoxy)acetate, tech
Catalog # APOSPC28111-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2,2-difluoro-2-(4-fluorophenoxy)acetate, tech
Catalog # APOSPC28111-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.145 g/mol
Storage Temperature:Ambient
CAS Number:338792-43-1
Formula:C₉H₇F₃O₃
MDL Number:MFCD00173165