Order Entry
Italy
ContactUsLinkComponent
1-Methyl-5-(morpholin-4-yl)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech
1-Methyl-5-(morpholin-4-yl)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech
Catalog # APOSPC28110-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-5-(morpholin-4-yl)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile, tech
Catalog # APOSPC28110-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.216 g/mol
Storage Temperature:Ambient
CAS Number:318517-82-7
Formula:C₁₀H₁₁F₃N₄O
MDL Number:MFCD00141044