5-[(2-Chloro-6-fluorophenyl)methoxy]-2-(hydroxymethyl)-4H-pyran-4-one, tech
Catalog # APOSPC28105-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.668 g/mol
Storage Temperature:Ambient
CAS Number:866040-48-4
Formula:C₁₃H₁₀ClFO₄
MDL Number:MFCD03787790