Order Entry
Italy
ContactUsLinkComponent
1-[2-(4-Fluorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
1-[2-(4-Fluorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
Catalog # APOSPC28104-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(4-Fluorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
Catalog # APOSPC28104-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.251 g/mol
Storage Temperature:Ambient
CAS Number:1226001-23-5
Formula:C₁₁H₈FNOS
MDL Number:MFCD16311596