Order Entry
Italy
ContactUsLinkComponent
6-Methyl-2-(morpholin-4-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile, tech
6-Methyl-2-(morpholin-4-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile, tech
Catalog # APOSPC28097-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-2-(morpholin-4-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile, tech
Catalog # APOSPC28097-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.238 g/mol
Storage Temperature:Ambient
CAS Number:478049-80-8
Formula:C₁₂H₁₂F₃N₃O
MDL Number:MFCD01316507