3-(Pyridin-3-yloxy)-5-(trifluoromethyl)pyridine-2-carbonitrile, tech
Catalog # APOSPC28086-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.191 g/mol
Storage Temperature:Ambient
CAS Number:338758-30-8
Formula:C₁₂H₆F₃N₃O
MDL Number:MFCD00664105