Order Entry
Italy
ContactUsLinkComponent
(3E)-4-(Benzylamino)-1,1,1-trifluorobut-3-en-2-one, tech
(3E)-4-(Benzylamino)-1,1,1-trifluorobut-3-en-2-one, tech
Catalog # APOSPC28074-1G
Supplier:  Apollo Scientific
CAS Number:  
(3E)-4-(Benzylamino)-1,1,1-trifluorobut-3-en-2-one, tech
Catalog # APOSPC28074-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.198 g/mol
Storage Temperature:Ambient
CAS Number:134219-70-8
Formula:C₁₁H₁₀F₃NO
MDL Number:MFCD01931068