Order Entry
Italy
ContactUsLinkComponent
7-[4-(3-Fluoropropoxy)phenyl]-[1,2,4]triazolo[1,5-a]pyrimidine, tech
7-[4-(3-Fluoropropoxy)phenyl]-[1,2,4]triazolo[1,5-a]pyrimidine, tech
Catalog # APOSPC28046-1G
Supplier:  Apollo Scientific
CAS Number:  
7-[4-(3-Fluoropropoxy)phenyl]-[1,2,4]triazolo[1,5-a]pyrimidine, tech
Catalog # APOSPC28046-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.278 g/mol
Storage Temperature:Ambient
CAS Number:478039-24-6
Formula:C₁₄H₁₃FN₄O
MDL Number:MFCD02571714