4-Fluorophenyl (2E)-3-(dimethylamino)prop-2-enoate, tech
Catalog # APOSPC28041-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:209.217 g/mol
Storage Temperature:Ambient
CAS Number:338404-21-0
Formula:C₁₁H₁₂FNO₂
MDL Number:MFCD00201970