Order Entry
Italy
ContactUsLinkComponent
3-[3-Fluoro-4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile, tech
3-[3-Fluoro-4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile, tech
Catalog # APOSPC28039-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[3-Fluoro-4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile, tech
Catalog # APOSPC28039-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.253 g/mol
Storage Temperature:Ambient
CAS Number:861211-70-3
Formula:C₁₃H₁₃FN₂O₂
MDL Number:MFCD06496173