Order Entry
Italy
ContactUsLinkComponent
2-(Morpholin-4-yl)-2-[3-(trifluoromethyl)phenyl]acetonitrile, tech
2-(Morpholin-4-yl)-2-[3-(trifluoromethyl)phenyl]acetonitrile, tech
Catalog # APOSPC27999-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Morpholin-4-yl)-2-[3-(trifluoromethyl)phenyl]acetonitrile, tech
Catalog # APOSPC27999-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.25 g/mol
Storage Temperature:Ambient
CAS Number:66548-59-2
Formula:C₁₃H₁₃F₃N₂O
MDL Number:MFCD00173086