Order Entry
Italy
ContactUsLinkComponent
2-Oxo-6-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
2-Oxo-6-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC27950-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Oxo-6-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC27950-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.203 g/mol
Storage Temperature:Ambient
CAS Number:140692-40-6
Formula:C₁₃H₇F₃N₂O
MDL Number:MFCD00215071