

4-Bromo-3-(trifluoromethyl)benzyl fluoride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2244084-02-2
:
Formula:C₈H₅BrF₄
MDL Number:MFCD31664906
:257.023 g/mol
UN:3265
ADR:8,II
:GHS05
:GHS08
Storage Temperature:Ambient
Frequently Bought Together