

3-Bromo-4-(trifluoromethoxy)benzyl fluoride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2169407-53-6
:
Formula:C₈H₅BrF₄O
MDL Number:MFCD31664895
:273.022 g/mol
UN:3265
ADR:8,III
:GHS05
:GHS08
Storage Temperature:Ambient
Frequently Bought Together