Order Entry
Italy
ContactUsLinkComponent
4-Bromophenyl 1,1,2,3,3,3-hexafluoropropyl ether 97%
Irritant
4-Bromophenyl 1,1,2,3,3,3-hexafluoropropyl ether 97%
Catalog # APOSPC2464-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromophenyl 1,1,2,3,3,3-hexafluoropropyl ether 97%
Catalog # APOSPC2464-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :323.03 g/mol
Boiling Pt:85 °C (15 mm Hg)
Density:1.7 g/cm³ (25 °C)
Flash Pt:107 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:52328-78-6
Formula:C₉H₅BrF₆O
MDL Number:MFCD04117777