3-(Trifluoromethyl)phenyl trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:199188-30-2
:
Formula:C₈H₄F₆O₃S
MDL Number:MFCD07784329
:294.171 g/mol
Storage Temperature:Ambient
Frequently Bought Together