Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Fluoro-4,6-dimethoxy-benzaldehyde ≥98%

2-Fluoro-4,6-dimethoxy-benzaldehyde ≥98%

Supplier:   Apollo Scientific
2-Fluoro-4,6-dimethoxy-benzaldehyde ≥98%
Supplier:  Apollo Scientific
Certificates
2-Fluoro-4,6-dimethoxy-benzaldehyde ≥98%

2 Options Available Below

Product Details & Documents
Synonyms:  2-Fluoro-4,6-dimethoxybenzaldehyde
  • CAS Number:214492-73-6
  • Formula:C₉H₉FO₃
  • MDL Number:MFCD18906188
  • Molecular Weight:184.164 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 2 040,00 €
    2 040,00 €
    1 * 100 mg
       2 040,00 €

  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 3 460,00 €
    3 460,00 €
    1 * 250 mg
       3 460,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.