[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1296225-26-7
:
Formula:C₆H₇F₃N₂O
MDL Number:MFCD15976765
:180.128 g/mol
Storage Temperature:Ambient
Frequently Bought Together