Ethyl 2-methyl-5-(((trifluoromethyl)-sulphonyl)-oxy)-1H-indole-3-carboxylate
Catalog # APOSPC210113-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:351.298 g/mol
Melting Pt:153…154 °C
Storage Temperature:Ambient
CAS Number:1796574-02-1
Formula:C₁₃H₁₂F₃NO₅S
MDL Number:MFCD28962696