Order Entry
Italy
ContactUsLinkComponent
8-chloro-3-phenyl-6-(trifluoromethyl)imidazo[1,5-a]pyridine
8-chloro-3-phenyl-6-(trifluoromethyl)imidazo[1,5-a]pyridine
Catalog # APOSPC210097-500MG
Supplier:  Apollo Scientific
CAS Number:  
8-chloro-3-phenyl-6-(trifluoromethyl)imidazo[1,5-a]pyridine
Catalog # APOSPC210097-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :296.675 g/mol
Storage Temperature:Ambient
CAS Number:1781241-49-3
Formula:C₁₄H₈ClF₃N₂
MDL Number:MFCD28805997