Order Entry
Italy
ContactUsLinkComponent
Methyl (2S)-2-acetamido-3-(2,3,6-trifluorophenyl)propanoate ≥ ee 80%
Methyl (2S)-2-acetamido-3-(2,3,6-trifluorophenyl)propanoate ≥ ee 80%
Catalog # APOSPC210068-500MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl (2S)-2-acetamido-3-(2,3,6-trifluorophenyl)propanoate ≥ ee 80%
Catalog # APOSPC210068-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :275.224 g/mol
Melting Pt:110…111 °C
Storage Temperature:Ambient
CAS Number:1630114-55-4
Formula:C₁₂H₁₂F₃NO₃
MDL Number:MFCD28127033