3-(4-Fluorophenyl)morpholine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1185045-95-7
:
Formula:C₁₀H₁₂FNO·ClH
MDL Number:MFCD03840090
:217.668 g/mol
Storage Temperature:Ambient
Frequently Bought Together