2,4-Dichloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053228-29-7
:
Formula:C₆H₂Cl₂FN₃
MDL Number:MFCD16249867
:206.005 g/mol
Storage Temperature:Ambient
Frequently Bought Together