Order Entry
Italy
ContactUsLinkComponent
2,3-Dichloro-4-(difluoromethyl)pyridine
2,3-Dichloro-4-(difluoromethyl)pyridine
Catalog # APOSPC200457-500MG
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dichloro-4-(difluoromethyl)pyridine
Catalog # APOSPC200457-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :197.998 g/mol
Storage Temperature:Ambient
CAS Number:1257071-85-4
Formula:C₆H₃Cl₂F₂N
MDL Number:MFCD22418352