Order Entry
Italy
ContactUsLinkComponent
Ethyl 3-cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylate
Ethyl 3-cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylate
Catalog # APOSPC200434-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylate
Catalog # APOSPC200434-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.197 g/mol
Melting Pt:57…59 °C
Storage Temperature:Ambient
CAS Number:1565827-82-8
Formula:C₁₁H₉F₃N₂O₂
MDL Number:MFCD27957302