Ethyl 3-amino-1-methyl-6-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1370587-21-5
:
Formula:C₁₂H₁₂F₃N₃O₂
MDL Number:MFCD21605748
:287.238 g/mol
Melting Pt:106…108 °C
Storage Temperature:Ambient
Frequently Bought Together