N-[4-(2-Fluoro-4-nitrophenoxy)pyridin-2-yl]morpholine-4-carboxamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:864245-86-3
:
Formula:C₁₆H₁₅FN₄O₅
MDL Number:MFCD22689174
:362.313 g/mol
Storage Temperature:Ambient
Frequently Bought Together