

2-(Trifluoromethyl)styrene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:395-45-9
:
Formula:C₉H₇F₃
MDL Number:MFCD00035984
:172.147 g/mol
Boiling Pt:61 °C
Density:1.164 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Refrigerator
EINECS:206-902-2
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together