Order Entry
Italy
ContactUsLinkComponent
N-(2,2-Dimethylpropanoyl)-4-(trifluoromethyl)aniline 98%
N-(2,2-Dimethylpropanoyl)-4-(trifluoromethyl)aniline 98%
Catalog # APOSPC1311-25G
Supplier:  Apollo Scientific
CAS Number:  
N-(2,2-Dimethylpropanoyl)-4-(trifluoromethyl)aniline 98%
Catalog # APOSPC1311-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :245.241 g/mol
Storage Temperature:Ambient
CAS Number:25617-34-9
Formula:C₁₂H₁₄F₃NO
MDL Number:MFCD03094126