

Perfluoro(3,7-dimethyloctanoic acid)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:172155-07-6
:
Formula:C₁₀HF₁₉O₂
MDL Number:MFCD01320738
:514.083 g/mol
Boiling Pt:95 °C (20 mm Hg)
UN:3265
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together