2-Morpholin-4-yl-2-[4-(trifluoromethyl)phenyl]ethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885950-67-4
:
Formula:C₁₃H₁₇F₃N₂O
MDL Number:MFCD01631947
:274.282 g/mol
Storage Temperature:Ambient
Frequently Bought Together