Order Entry
Italy
ContactUsLinkComponent
2'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde 95+%
2'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde 95+%
Catalog # APOSPC11131-1G
Supplier:  Apollo Scientific
CAS Number:  
2'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde 95+%
Catalog # APOSPC11131-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.208 g/mol
Storage Temperature:Ambient
CAS Number:57592-42-4
Formula:C₁₃H₉FO
MDL Number:MFCD03424638