Ethyl 5,7-difluoro-4-hydroxyquinoline-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:228728-82-3
:
Formula:C₁₂H₉F₂NO₃
MDL Number:MFCD00173415
:253.202 g/mol
Storage Temperature:Ambient
Frequently Bought Together