
4,8-Dichloro-5-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:959071-80-8
:
Formula:C₁₀H₄Cl₂F₃N
MDL Number:MFCD28010391
:266.047 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together