[Difluoro(phenylsulphanyl)methyl]trimethylsilane 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:536975-49-2
:
Formula:C₁₀H₁₄F₂SSi
MDL Number:MFCD19442557
:232.365 g/mol
Storage Temperature:Ambient
Frequently Bought Together