Order Entry
Italy
ContactUsLinkComponent
(E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)acrylic acid 95%
Irritant
(E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)acrylic acid 95%
Catalog # APOSPC0880-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(E)-3-(2,2-Difluoro-1,3-benzodioxol-5-yl)acrylic acid 95%
Catalog # APOSPC0880-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.149 g/mol
Melting Pt:176…179 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:387350-55-2
Formula:C₁₀H₆F₂O₄
MDL Number:MFCD00203111