1,2-Bis(4-fluorophenyl)ethane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:458-76-4
:
Formula:C₁₄H₁₂F₂
MDL Number:MFCD00153123
Boiling Pt:269 °C
Melting Pt:90 °C
:218.242 g/mol
Storage Temperature:Ambient
Frequently Bought Together