
5-Nitro-2-[3-(trifluoromethyl)phenoxy]benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:217186-14-6
:
Formula:C₁₄H₈F₃NO₄
MDL Number:MFCD03001331
:311.213 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together