

1-Bromo-1,1,2,2-tetrafluoropentane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:151831-44-6
:
Formula:C₅H₇BrF₄
MDL Number:MFCD00153716
:223.007 g/mol
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together