Order Entry
Italy
ContactUsLinkComponent
 
Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
  APOSPC0541-1G
 :  
Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
  APOSPC0541-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :222.164 g/mol
 :58…61 °C
 :GHS07
 :Ambient
 :231285-86-2
 :C₈H₉F₃N₂O₂
 :MFCD00277430