
8-Methoxy-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine 97%
Catalog # APOSPC0235-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:294.232 g/mol
Melting Pt:142 °C
:GHS07
Storage Temperature:Ambient
CAS Number:230295-08-6
Formula:C₁₃H₉F₃N₄O
MDL Number:MFCD01073588