
Heptafluoroisopropyl 4-iodo-1,1,2,2-tetrafluorobut-1-yl ether 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25080-20-0
:
Formula:C₇H₄F₁₁IO
MDL Number:MFCD23381072
:439.993 g/mol
Boiling Pt:85…87 °C (100 mm Hg)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together