
2,2-Difluoropropane-1,3-diol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:428-63-7
:
Formula:C₃H₆F₂O₂
MDL Number:MFCD16619633
:112.075 g/mol
Melting Pt:51…52 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together